Learn More
Novus Biologicals™ Lightning-Link (R) R-PE Antibody Labeling Kit
Código de producto. p-7070036 Tienda Bio Techne Productos
Click to view available options
Cantidad:
3 reacciones de 0,01 mg
3 reacciones de 60 μg
5 reacciones de 1 mg
Reacción de 1 x 100 μg
Reacción de 1 x 5 mg
Reacción de 1 mg
Tamaño de la unidad:
1 set
3 miligramos
60 microgramos
600 microgramos
Código de producto. 18785583
Marca: Novus Biologicals™ 7030003
Applications: Flow Cytometry
R-Phycoerythrin (R-PE) is a fluorescent protein from the phycobiliprotein family, and is isolated from red algae. The absorbance spectrum of R-PE covers a broad range of excitation wavelengths, which provides an advantage for multi-color immunofluorescent staining or cell sorting. R-PE is one of the most intensely fluorescent phycobiliprotein having orange fluorescence. It is significantly brighter and more photostable than conventional organic fluorophores and it has a high quantum yield.
Información de órdenes
Cantidad
TRUSTED_SUSTAINABILITY
Especificaciones
Contenido y almacenamiento Store at -20°C in the dark. Avoid freeze-thaw cycles.
Cantidad 5 reacciones de 1 mg
Conjugado PE
Tipo de producto R-PE Antibody Labeling Kit