missing translation for 'onlineSavingsMsg'
Learn More

LIFR alpha Antibody (32953), DyLight 350, Novus Biologicals™

Product Code. 18014865 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.5 mL
Unit Size:
0.50 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18014865 0.5 mL 0.50 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18014865 Brand Novus Biologicals Brand No. FAB249D

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LIFR alpha Monoclonal antibody specifically detects LIFR alpha in Human samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno LIFR alpha
Aplicaciones Western Blot, Flow Cytometry
Clasificación Monoclonal
Clon 32953
Conjugado DyLight 350
Formulación 50mM Sodium Borate
Alias de gen CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
Especie del huésped Mouse
Inmunógeno S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LIF R alpha , Gln45-Ser833, Accession # P42702
Método de purificación Protein A or G purified from ascites
Cantidad 0.5 mL
Estado normativo RUO
Disciplina de investigación Immunology
Primario o secundario Primary
ID de gen (Entrez) 3977
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.