missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LGTN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12840-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a translation initiation factor involved in the recruitment and delivery of aminoacyl-tRNAs to the P-site of the eukaryotic ribosome in a GTP-independent manner. This gene was previously referred to as ligatin, but is now known to localize to the cytoplasm and localize and function with translation factors. Multiple transcript variants encoding different isoforms have been found for this gene.Especificaciones
| LGTN | |
| Polyclonal | |
| Unconjugated | |
| EIF2D | |
| EIF2D, HCA56, LGTN, ligatin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16865, 1939, 498225 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P41214, Q5PPG7, Q61211 | |
| Eif2d | |
| LGTN Fusion Protein Ag3579 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido