Learn More
Lgr5/GPR49 Antibody (CL13579) [Alexa Fluor™ 647], Novus Biologicals™
Código de producto. 30308441 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30308441 0.1 mL 0.10 ml
1 options
Código de producto. 30308441 Proveedor Novus Biologicals N.º de proveedor NBP324571AF647
Mouse Monoclonal Antibody
Lgr5/GPR49 Monoclonal antibody specifically detects Lgr5/GPR49 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Lgr5/GPR49
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL13579
Conjugado Alexa Fluor 647
Formulación 50mM Sodium Borate
Alias de gen FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from O75473, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway
Primario o secundario Primary
ID de gen (Entrez) 8549
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1