missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lewis Y Mouse, Clone: LWY/1463, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio MSM3-1463-P0
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Lewis Y | |
Cell membrane, cytoplasmic and extracellular | |
LWY/1463 | |
Unconjugated | |
Mouse | |
Lung, Colon, Bladder or Hepatocellular Carcinoma | |
20 μg | |
Primary | |
IgG1 κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
Human colon carcinoma cells | |
Protein A/G Purified | |
RUO | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |