missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leukotriene B4 R1 Antibody (202/7B1), Allophycocyanin/Cy7, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Leukotriene B4 R1 Monoclonal specifically detects Leukotriene B4 R1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antígeno | Leukotriene B4 R1 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | 202/7B1 |
| Conjugado | APC-Cyanine7 |
| Dilución | Flow Cytometry |
| Formulación | PBS with 0.05% Sodium Azide |
| Alias de gen | chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 |
| Símbolos de los genes | LTB4R |
| Especie del huésped | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?