missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Leptin Recombinant Protein

Código de producto. 11534930
Change view
Click to view available options
Cantidad:
1 mg
Tamaño de la unidad:
1 miligramo
Código de producto. Cantidad unitSize
11534930 1 mg 1 miligramo
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11534930

Marca: Invitrogen™ RP10913

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

This is a recombinant protein suitable for use as a control. This protein is soluble in water and most aqueous buffers above and below the isoelectric point.

Leptin is a 167 amino acid long protein hormone with important effects in regulating body weight, metabolism and reproductive function. Leptin is approximately 16 kDa in mass and encoded by the obese (ob) gene. Leptin is expressed predominantly by adipocytes, which fits with the idea that body weight is sensed as the total mass of fat in the body. Smaller amounts of leptin are also secreted by cells in the epithelium of the stomach and in the placenta. Leptin receptors are highly expressed in areas of the hypothalamus known to be important in regulating body weight, as well as in T lymphocytes and vascular endothelial cells. Leptin's effects on body weight are mediated through effects on hypothalamic centers that control feeding behavior and hunger, body temperature and energy expenditure. Leptin is involved in regulating food intake, energy expenditure, and adiposity through hypothalamic leptin receptors. Leptin promotes hematopoiesis, angiogenesis, wound healing, inflammation, immune responses, influences pubertal development and fetal growth. Studies have investigated the role of leptin in obesity, anorexia nervosa, insulin resistance, and hypertension. Leptin also has thermogenic actions and regulates enzymes of fatty acid oxidation. Severe hereditary obesity in rodents and humans can be caused by defects in leptin production.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P41159
Para utilizar con (aplicación) Control
Formulación 45 mM sodium bicarbonate with no preservative
ID de gen (Entrez) 3952
Peso molecular 16.2 kDa
Nombre Human Leptin
Cantidad 1 mg
Fuente E. Coli
Inmunógeno N/A
Estado normativo RUO
Alias de gen CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin
Nombre común Leptin
Símbolo de gen LEP
Actividad biológica ED50 = 0.5 - 1.6 ng/mL; determined by the dose-dependent stimulation of human OB-R transfected BaF3 indicator cells.
Conjugado Unconjugated
Especie E. coli
Recombinante Recombinant
Secuencia Human Leptin recombinant protein contains 147 amino acids
Contenido y almacenamiento -20°C, Avoid Freeze/Thaw Cycles
Sistema de expresión E. coli
Formulario Lyophilized
Subtipo de proteína Recombinant
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.