missing translation for 'onlineSavingsMsg'
Learn More

LENG1 Antibody (OTI10A7) - Azide and BSA Free, Novus Biologicals™

Código de producto. 18199543 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18199543 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18199543 Proveedor Novus Biologicals N.º de proveedor NBP272202

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

LENG1 Monoclonal specifically detects LENG1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno LENG1
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, CyTOF
Clasificación Monoclonal
Clon OTI10A7
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 1:500, Flow Cytometry 1:100, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100, CyTOF-ready
Formulación Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Alias de gen leukocyte receptor cluster (LRC) member 1, leukocyte receptor cluster member 1
Símbolos de los genes LENG1
Especie del huésped Mouse
Inmunógeno Full length human recombinant protein of human LENG1 (NP_077292) produced in HEK293T cell.
Peso molecular del antígeno 30.3 kDa
Método de purificación Affinity Purified
Cantidad 100 μg
Primario o secundario Primary
ID de gen (Entrez) 79165
Reconstitución Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Especies diana Human
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulario Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.