missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Lefty-1 Polyclonal specifically detects Lefty-1 in Human samples. It is validated for Western Blot.
Especificaciones
Especificaciones
| Antígeno | Lefty-B |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Concentración | 1mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:3000 |
| Formulación | 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal |
| N.º de referencia del gen | O75610 |
| Alias de gen | LEFTBProtein lefty-B, left-right determination factor 1, Left-right determination factor B, left-right determination, factor B, LEFTYBProtein lefty-1 |
| Símbolos de los genes | LEFTY1 |
| Mostrar más |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?