missing translation for 'onlineSavingsMsg'
Learn More

LC3B Antibody - BSA Free, Novus Biologicals™

Código de producto. p-5078400 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
Código de producto. Cantidad unitSize
18760174 0.025 mL 25 microlitros
18795583 0.1 mL 0.10 ml
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18760174

Marca: Novus Biologicals NB6001384SS

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody has been used in 316 publications

LC3B Polyclonal specifically detects LC3B in Human, Mouse, Rat, Porcine, Bacteria, Bovine, Canine, Primate, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Electron Microscopy, Immunohistochemistry Free-Floating, Knockout Validated, Knockdown Validated.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno LC3B
Aplicaciones Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Immunoassay, Immunohistochemistry (Free Floating), KnockDown
Clasificación Polyclonal
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:1000, Simple Western 1:100, Flow Cytometry 1:200, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 0.1-2 ug/ml, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:400, Immunohistochemistry-Frozen 1:400, Immunoblotting, Electron Microscopy, Immunohistochemistry Free-Floating, Knockout Validated, Knockdown Validated
Formulación PBS with 0.02% Sodium Azide
N.º de referencia del gen Q9GZQ8
Alias de gen ATG8F, Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B
Símbolos de los genes MAP1LC3B
Especie del huésped Rabbit
Inmunógeno Polyclonal LC3B Antibody was made to a synthetic peptide made to the N-terminal region of the human LC3B protein. [Uniprot: Q9GZQ8]
Peso molecular del antígeno 14.688 kDa
Método de purificación Affinity Purified
Cantidad 0.025 mL
Estado normativo RUO
Disciplina de investigación Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Hypoxia, Innate Immunity, Lipid and Metabolism, Macroautophagy, Mitophagy, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 81631
Especificidad de la prueba Human, mouse, rat and bovine. Zebrafish reactivity reported in scientific literature (PMID: 23724125). Canine and primate reactivity reported in scientific literature (PMID: 24027311) Porcine reactivity reported in scientific literature (PMID: 25378587) Other species have not been tested.
Especies diana Human, Mouse, Rat, Pig, Bacteria, Bovine, Canine, Primate, Zebrafish
Contenido y almacenamiento Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.