Learn More
LC3 Antibody Pack, Novus Biologicals™
Código de producto. 18548901 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 Pack
Tamaño de la unidad:
1 set
18548901 1 Pack 1 set
1 options
Código de producto. 18548901 Proveedor Novus Biologicals N.º de proveedor NB910407521Pack
Antibody Pack
LC3 Antibody Packantibody specifically detects LC3 in Human, Mouse, Rat, Porcine, Bacteria, Bovine, Canine, Primate, Zebrafish samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, ChIP assay (ChIP), Gene Knock-Out, KnockDown
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno LC3
Aplicaciones Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry
Conjugado Unconjugated
Alias de gen ATG8F, LC3B, MAP1A/1BLC3, map1lc3b, MAP1LC3B-a, microtubule associated protein 1 light chain 3 beta
Inmunógeno For NB600-1384: Polyclonal LC3B Antibody was made to a synthetic peptide made to the N-terminal region of the human LC3B protein [Uniprot: Q9GZQ8]., For NB100-2331: A synthetic peptide made to an internal portion of the human LC3 protein sequence (between residues 25-121) [Uniprot: Q9H492]., For NB100-2220: Polyclonal LC3B Antibody was made to a synthetic peptide made to an N-terminal portion of the human LC3B protein sequence (between residues 1-100) [UniProt# Q9GZQ8].
Cantidad 1 Pack
Estado normativo RUO
Disciplina de investigación Autophagy, Cancer, Cardiovascular Biology, mTOR Pathway
ID de gen (Entrez) 81631
Especies diana Human, Mouse, Rat, Pig, Bacteria
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.