Learn More
Piercan™ Latex Dry Gloves
Formulated from Natural Rubber (polyisoprene). Piercan™ Latex Dry Gloves react well to acids, dilute bases and alcohols. The gloves are resistant to cold conditions and provide good flexibility and dexterity.
85.60€ - 179.00€
Especificaciones
Color | Natural |
---|---|
Desechables | Single-use |
Hand | Ambidextrous |
Material | Latex |
Guantes con puño de color | Natural |
Código de producto | Marca | Tamaño | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Tamaño | Precio | Cantidad y disponibilidad | |||||
11811250
|
Piercan™
000393 10800 H 8 6/10 |
8 | |||||||
11727108
|
Piercan™
000399 10800 H 10 6/10 |
10 | |||||||
11831250
|
Piercan™
13800 H 7 E6 |
7 | |||||||
11801260
|
Piercan™
18800 H 9 E4 |
9 | |||||||
11811260
|
Piercan™
97800 H 9 E4 |
9 | |||||||
11737108
|
Piercan™
18800 H 8 E4 |
8 | |||||||
Descripción
- Glove manufactured in natural rubber (polyisoprene)
- Packing by pair inside a black pouch
- Can be delivered decontaminated or decontaminated-sterile (Gamma-ray sterilization)
- Classified as non-toxic waste providing they have not been contaminated during use
Especificaciones
Natural | |
Ambidextrous | |
Natural |
Single-use | |
Latex |