Learn More
LAPTM4B Antibody (CL5289) [Alexa Fluor™ 594], Novus Biologicals™
Código de producto. 30303020 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30303020 0.1 mL 0.10 ml
1 options
Código de producto. 30303020 Proveedor Novus Biologicals N.º de proveedor NBP261158AF594
Mouse Monoclonal Antibody
LAPTM4B Monoclonal antibody specifically detects LAPTM4B in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno LAPTM4B
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Clasificación Monoclonal
Clon CL5289
Conjugado Alexa Fluor 594
Formulación 50mM Sodium Borate
Alias de gen LAPTM4beta, LC27, lysosomal associated protein transmembrane 4 beta, lysosomal protein transmembrane 4 beta, lysosomal-associated transmembrane protein 4B, Lysosome-associated transmembrane protein 4-beta
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from Q86VI4, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 55353
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1