Learn More
LAPTM4B Antibody (CL5287) [Allophycocyanin], Novus Biologicals™
Código de producto. 30302575 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30302575 0.1 mL 0.10 ml
1 options
Código de producto. 30302575
Marca: Novus Biologicals NBP261157APC
Mouse Monoclonal Antibody
LAPTM4B Monoclonal antibody specifically detects LAPTM4B in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno LAPTM4B
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL5287
Conjugado APC
Formulación PBS
Alias de gen LAPTM4beta, LC27, lysosomal associated protein transmembrane 4 beta, lysosomal protein transmembrane 4 beta, lysosomal-associated transmembrane protein 4B, Lysosome-associated transmembrane protein 4-beta
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from Q86VI4, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 55353
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG2b