Learn More
LAP (TGF-beta 1) Antibody (1015022), Allophycocyanin, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
LAP (TGF-beta 1) Monoclonal antibody specifically detects LAP (TGF-beta 1) in Human samples. It is validated for ELISA, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Especificaciones
Especificaciones
| Antígeno | LAP (TGF-beta 1) |
| Aplicaciones | ELISA, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 1015022 |
| Conjugado | APC |
| Dilución | ELISA, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready |
| Formulación | PBS |
| Alias de gen | CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor beta 1 |
| Especie del huésped | Mouse |
| Inmunógeno | Chinese hamster ovary cell line CHO-derived recombinant human LAP (TGF-beta 1), Met1-Ser390, Accession # P01137 |
| Mostrar más |