Learn More
Langerin/CD207 Antibody (LGRN/7430), Novus Biologicals™
Código de producto. 18607924 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
20 μg
Tamaño de la unidad:
100 microgramos
20 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18607924 100 μg 100 microgramos
18603545 20 μg 20 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18607924 Proveedor Novus Biologicals N.º de proveedor NBP320556100ug
Mouse Monoclonal Antibody
Langerin/CD207 Monoclonal antibody specifically detects Langerin/CD207 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Langerin/CD207
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon LGRN/7430
Concentración 0.2 mg/ml
Conjugado Unconjugated
Dilución Immunohistochemistry-Paraffin 1-2 μg/mL
Formulación 10mM PBS with 0.05% BSA
Alias de gen CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin
Especie del huésped Mouse
Inmunógeno Recombinant fragment (around aa50-250) of human Langerin/CD207 protein (exact sequence is proprietary)
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación metabolism, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 50489
Especies diana Human
Contenido y almacenamiento Store at 4&drg;C. Do not freeze.
Formulario Purified
Isotype IgG