Learn More
LAMP-1/CD107a Antibody (CL4472) [Janelia Fluor™ 525], Novus Biologicals™
Código de producto. 30294562 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30294562 0.1 mL 0.10 ml
1 options
Código de producto. 30294562 Proveedor Novus Biologicals N.º de proveedor NBP259052JF525
Mouse Monoclonal Antibody
LAMP-1/CD107a Monoclonal antibody specifically detects LAMP-1/CD107a in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno LAMP-1/CD107a
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL4472
Conjugado Janelia Fluor 525
Formulación 50mM Sodium Borate
Alias de gen CD107 antigen-like family member A, CD107a, CD107a antigen, LAMP-1, LAMPA, LGP120, lysosomal-associated membrane protein 1, lysosome-associated membrane glycoprotein 1, Lysosome-associated membrane protein 1
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from P11279, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Autophagy, Cholesterol Metabolism, Endosome Markers, Glycobiology, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Virology Bacteria and Parasites
Primario o secundario Primary
ID de gen (Entrez) 3916
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1