missing translation for 'onlineSavingsMsg'
Learn More

Laminin S/Laminin beta 2 Antibody (CL2976) [Janelia Fluor™ 525], Novus Biologicals™

Código de producto. 30286986 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30286986 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30286986 Proveedor Novus Biologicals N.º de proveedor NBP242386JF525

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Mouse Monoclonal Antibody

Laminin S/Laminin beta 2 Monoclonal antibody specifically detects Laminin S/Laminin beta 2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Laminin S/Laminin beta 2
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
Clasificación Monoclonal
Clon CL2976
Conjugado Janelia Fluor 525
Formulación 50mM Sodium Borate
Alias de gen Laminin B1s chain, laminin S, laminin subunit beta-2, laminin, beta 2 (laminin S), Laminin-11 subunit beta, Laminin-14 subunit beta, Laminin-15 subunit beta, Laminin-3 subunit beta, Laminin-4 subunit beta, Laminin-7 subunit beta, LAMSLaminin-9 subunit beta, S-LAM beta, S-laminin subunit beta
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P55268, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3913
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.