missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Lamin A + C Polyclonal specifically detects Lamin A + C in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antígeno | Lamin A + C |
| Aplicaciones | Western Blot, Immunocytochemistry, Immunofluorescence |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:1000 |
| Formulación | Supplied as concentrated IgY in PBS with 5mM Sodium Azide |
| N.º de referencia del gen | P02545 |
| Alias de gen | CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32 |
| Símbolos de los genes | LMNA |
| Especie del huésped | Chicken |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?