Descripción
LAG-3 Monoclonal antibody specifically detects LAG-3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry
Especificaciones
Especificaciones
| Antígeno | LAG-3 |
| Aplicaciones | Western Blot, Flow Cytometry, Immunohistochemistry |
| Clasificación | Monoclonal |
| Conjugado | Unconjugated |
| Formulación | See individual datasheets |
| Alias de gen | 17b4, 17b4 lag3, 17b4 neutralizing, anti human lag3, Anti-LAG-3, CD223, CD223 Antigen, Human LAG3, Human LAG-3, LAG3, LAG-3, LAG-3 17b4, LAG-3 blocking, lymphocyte activating 3, lymphocyte activation gene 3 protein, lymphocyte-activation gene 3, Secreted lymphocyte activation gene 3 protein, sLAG-3 |
| Inmunógeno | See individual datasheets |
| Cantidad | 3 Vials |
| Estado normativo | RUO |
| Disciplina de investigación | Adaptive Immunity, Cancer, Immunology, Innate Immunity |
| Mostrar más |