Learn More
Epredia™ Lab Vision™ S100 A6 Ab-9 Rabbit Polyclonal Antibody with BSA and azide
Epredia™ S100 A6 Ab-9, Rabbit Polyclonal Antibody provides accurate, reproducible results.
Marca: Epredia™ RB-1805-A
Descripción
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an α and β chain whereas S100B is composed of two β chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant human S100 A6 monomer expressed in E. coli
Molecular Weight: 10-12kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Especificaciones
| S100 A6 Ab-9 | |
| Polyclonal | |
| Rabbit | |
| Tonsil | |
| RUO | |
| Primary | |
| Concentrated |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Purified recombinant human S100 A6 monomer expressed in E. coli | |
| 1 mL | |
| Neuroscience | |
| Bovine, Canine, Equine, Feline, Human, Mouse | |
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.