missing translation for 'onlineSavingsMsg'
Learn More

KIR2DS5/CD158g Antibody (1165A) [Biotin], Novus Biologicals™

Product Code. 18607774 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
18607774 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 18607774

Brand: Novus Biologicals FAB8920B

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

KIR2DS5/CD158g Monoclonal antibody specifically detects KIR2DS5/CD158g in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno KIR2DS5/CD158g
Aplicaciones Flow Cytometry
Clasificación Monoclonal
Clon 1165A
Conjugado Biotin
Dilución Flow Cytometry
Formulación PBS
Alias de gen killer cell immunoglobulin-like receptor 2DS5, CD158G, killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 5, NKAT9
Especie del huésped Rabbit
Inmunógeno NS0 mouse myeloma cell line transfected with human KIR2DS5/CD158g, His22-His245, Accession # Q14953
Método de purificación Protein A or G purified from cell culture supernatant
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3810
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.