missing translation for 'onlineSavingsMsg'
Learn More

Kininogen 1 Antibody, Novus Biologicals™

Product Code. 18642869 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Unit Size:
100 microlitros
50 microlitros
Product Code. Cantidad unitSize
18642869 100 μg 100 microlitros
18635857 50 μg 50 microlitros
2 options
This item is not returnable. View return policy

Product Code. 18642869

Brand: Novus Biologicals NBP299365100ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Kininogen 1 Polyclonal antibody specifically detects Kininogen 1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Kininogen 1
Aplicaciones ELISA
Clasificación Polyclonal
Conjugado Unconjugated
Dilución ELISA 1:5000-1:10000
Formulación 0.2 um filtered solution in PBS
Alias de gen Alpha-2-thiol proteinase inhibitor, BDK, BK, bradykinin, Fitzgerald factor, High molecular weight kininogen, HMWK, kininogen-1, Williams-Fitzgerald-Flaujeac factor
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with purified, recombinant Human Kininogen 1 (Accession#: NP_001095886.1; Gln19-Ser644)
Método de purificación Antigen and protein A Affinity-purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Apoptosis, Immunology
Primario o secundario Primary
ID de gen (Entrez) 3827
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.