missing translation for 'onlineSavingsMsg'
Learn More

KIF2C Antibody (KIF2C/4705), Novus Biologicals™

Código de producto. 18692564 Tienda Bio Techne Productos
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
100 μg
20 μg
missing translation for 'unitSize'
100 microgramos
20 microgramos
Product Code. Cantidad unitSize
18692564 20 μg 20 microgramos
18630644 100 μg 100 microgramos
2 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 18692564

missing translation for 'mfr': Novus Biologicals NBP32054020ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KIF2C Monoclonal antibody specifically detects KIF2C in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno KIF2C
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon KIF2C/4705
Concentración 0.2 mg/ml
Conjugado Unconjugated
Dilución Immunohistochemistry-Paraffin 1-2 μg/mL
Formulación 10mM PBS with 0.05% BSA
Alias de gen kinesin family member 2C, kinesin-like 6 (mitotic centromere-associated kinesin), Kinesin-like protein 6, kinesin-like protein KIF2C, MCAKKNSL6kinesin-like 6, Mitotic centromere-associated kinesin
Especie del huésped Mouse
Inmunógeno Recombinant fragment (around aa500-700) of human KIF2C protein (exact sequence is proprietary)
Método de purificación Protein A or G purified
Cantidad 20 μg
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Cellular Markers, DNA Repair
Primario o secundario Primary
ID de gen (Entrez) 11004
Especies diana Human
Contenido y almacenamiento Store at 4&drg;C. Do not freeze.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.