missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIAA0087 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24973-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Description
This KIAA0087 antibody gives a 40 kDa band in Western Blot.
KIAA0087 (KIAA0087 LncRNA) is an RNA Gene, and is affiliated with the non-coding RNA class. Diseases associated with KIAA0087 include Endometrial Cancer and Pancreatic Cancer.Spécification
| KIAA0087 | |
| Polyclonal | |
| Unconjugated | |
| KIAA0087 | |
| KIAA0087 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 9808 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14695 | |
| KIAA0087 | |
| KIAA0087 Fusion Protein Ag21759 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu