missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Keratin 31 Monoclonal specifically detects Keratin 31 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
Especificaciones
| Antígeno | Keratin 31 |
| Aplicaciones | Western Blot, Immunohistochemistry |
| Clasificación | Monoclonal |
| Clon | LHTric-1 |
| Concentración | 1.0 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot, Immunohistochemistry |
| Formulación | PBS with 0.02% Sodium Azide |
| Alias de gen | Hair Keratin, Type I Ha, Hard Keratin, Type I, 1, hHa1, K31, Keratin 31, Keratin, Hair, Acidic, 1, Keratin, Type I Cuticular Ha1, Keratin-31, KRTHA1 |
| Símbolos de los genes | KRT31 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?