missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human KDR (VEGFR2), His Tag Recombinant Protein

Código de producto. 11523497
Click to view available options
Cantidad:
100 μg
10 μg
Tamaño de la unidad:
10 microgramos
100 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 11523497

Marca: Invitrogen™ PV3660

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic.

KDR (FLK1, VEGFR2) is a type III receptor tyrosine kinase. It binds vascular endothelial growth factor with high affinity in vitro, and is expressed early in development by endothelial cell precursors. FLK1 (VEGF Receptor 2) encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc. Mutations of FLK1 (VEGF Receptor 2) are implicated in infantile capillary hemangiomas.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P35968
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50mM tris with 2mM DTT, 0.25mM EDTA, 50% glycerol, 150mM NaCl, 0.01% Triton X-100 and no preservative; pH 7.5
ID de gen (Entrez) 3791
Peso molecular 68.4 kDa
Nombre Human KDR (VEGFR2), His Tag
Intervalo de pH 7.5
Método de purificación Purified
Cantidad 10 μg
Requisitos de almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen 6130401C07; CD309; CD309 antigen; fetal liver kinase 1; fetal liver kinase-1; FLK1; Flk-1; FLK1 kinase insert domain receptor (a type III receptor tyrosine kinase) (VEGF receptor 2); FLK1 kinase insert domain receptor (VEGF receptor 2); flk-1 type VEGF receptor; KDR; kinase insert domain protein receptor; kinase insert domain receptor; kinase insert domain receptor (a type III receptor tyrosine kinase); kinase NYK; Krd-1; Ly73; protein-tyrosine kinase receptor flk-1; sCD309; soluble CD309; soluble vascular endothelial growth factor receptor 2; soluble VEGFR 2; soluble VEGFR2; sVEGFR-2; tyrosine kinase growth factor receptor; vascular endotheial growth factor 2; vascular endothelial growth factor receptor 2; vascular endothelial growth factor receptor- 2; vascular endothelial growth factor receptor-2; vascular endothelial growth factor receptor-3; VEGF R2; VEGF receptor-2; VEGFR; VEGFR2; VEGFR-2
Nombre común FLK1 (VEGF Receptor 2)
Símbolo de gen KDR
Familia Quinasa VEGF Receptors
Longitud de la proteína 789-1356
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína His-tag
Secuencia 789-1356
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Entrada del ensayo Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic
Grupo de cinasas Tyrosine Kinases
Condiciones de envío Dry Ice
Compatibilidad de sistemas LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Entrada diana KDR
Gen diana kinase insert domain receptor
Sistema de expresión Insect cells
Familia de proteínas Kinases & Inhibitors
Forma de proteína Recombinant, Cytoplasmic
Formulario Liquid
Subtipo de proteína Receptor Tyrosine Kinases
Categoría de investigación Angiogenesis, Signal Transduction
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado