missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNIP4 Mouse anti-Human, Porcine, Rat, Clone: 5B1D12, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 60133-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
KCNIP4 encodes a member of the family of voltage-gated potassium (Kv) channel-interacting proteins (KCNIPs), which belong to the recoverin branch of the EF-hand superfamily. Members of the KCNIP family are small calcium binding proteins. They all have EF-hand-like domains, and differ from each other in the N-terminus. They are integral subunit components of native Kv4 channel complexes. They may regulate A-type currents, and hence neuronal excitability, in response to changes in intracellular calcium. This protein member also interacts with presenilin.Especificaciones
KCNIP4 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PIL6, Q99MG9 | |
KCNIP4 | |
KCNIP4 Fusion Protein Ag4863 | |
150 μL | |
Primary | |
Human, Porcine, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
5B1D12 | |
Unconjugated | |
KCNIP4 | |
CALP, Calsenilin like protein, KCHIP4, KCNIP4 | |
Mouse | |
Caprylic Acid Precipitation | |
RUO | |
100516499, 259243, 80333 | |
-20°C | |
Liquid |