missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNE4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18289-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
Ancillary protein that assembles as a beta subunit with a voltage-gated potassium channel complex of pore-forming alpha subunits. Modulates the gating kinetics and enhances stability of the channel complex. May associate with KCNQ1/KVLTQ1 and inhibit potassium current.Especificaciones
KCNE4 | |
Polyclonal | |
Unconjugated | |
KCNE4 | |
KCNE4, MinK related peptide 3, MIRP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23704, 367302, 57814 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8WWG9, Q9WTW3 | |
Kcne4 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |