missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAP1 Mouse anti-Human, Clone: 1B9G12, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66630-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TIF1 beta is a member of the tripartite motif (TRIM) family. It is composed of a N-terminal RBCC (RING finger, B boxes, coiled coil) motif, and a C-terminal bromodomain which is preceded by a PHD finger. TIF1 beta has been reported to function as a co-repressor for Kruppel-associated box (KRAB)-domain-containing zinc finger proteins. As such, it plays a crucial role in early embryogenesis. It appears that the co-repressor function of TIF1 beta is related to its organization and/or maintece of higher-order, heterochromatin-like structures.Especificaciones
| KAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13263 | |
| TRIM28 | |
| KAP1 Fusion Protein Ag7519 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 1B9G12 | |
| Unconjugated | |
| TRIM28 | |
| KAP 1, KAP1, KRAB associated protein 1, KRAB interacting protein 1, KRIP 1, Nuclear corepressor KAP 1, RING finger protein 96, RNF96, TF1B, TIF1 beta, TIF1B, TRIM28, tripartite motif containing 28 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10155 | |
| -20°C | |
| Liquid |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?