missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAP1 Mouse anti-Human, Clone: 1B9G12, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66630-1-IG-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TIF1 beta is a member of the tripartite motif (TRIM) family. It is composed of a N-terminal RBCC (RING finger, B boxes, coiled coil) motif, and a C-terminal bromodomain which is preceded by a PHD finger. TIF1 beta has been reported to function as a co-repressor for Kruppel-associated box (KRAB)-domain-containing zinc finger proteins. As such, it plays a crucial role in early embryogenesis. It appears that the co-repressor function of TIF1 beta is related to its organization and/or maintece of higher-order, heterochromatin-like structures.Especificaciones
| KAP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13263 | |
| TRIM28 | |
| KAP1 Fusion Protein Ag7519 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1B9G12 | |
| Unconjugated | |
| TRIM28 | |
| KAP 1, KAP1, KRAB associated protein 1, KRAB interacting protein 1, KRIP 1, Nuclear corepressor KAP 1, RING finger protein 96, RNF96, TF1B, TIF1 beta, TIF1B, TRIM28, tripartite motif containing 28 | |
| Mouse | |
| Protein A | |
| RUO | |
| 10155 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido