missing translation for 'onlineSavingsMsg'
Learn More

KAP1 Antibody, HRP, Novus Biologicals™

Código de producto. 18594923 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
18594923 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18594923

Marca: Novus Biologicals NB500158H

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

KAP1 Polyclonal antibody specifically detects KAP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno KAP1
Aplicaciones Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado HRP
Dilución Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulación PBS
Alias de gen E3 SUMO-protein ligase TRIM28, EC 6.3.2.-, FLJ29029, KAP-1, KAP1KRAB-associated protein 1, KRIP-1, Nuclear corepressor KAP-1, RNF96KRAB-interacting protein 1, TF1B, TIF1-beta, TIF1BRING finger protein 96, transcription intermediary factor 1-beta, transcriptional intermediary factor 1-beta, tripartite motif containing 28, tripartite motif-containing 28, Tripartite motif-containing protein 28
Especie del huésped Rabbit
Inmunógeno The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human Tripartite Motif-Containing 28 using the numbering given in entry NP_005753.1 (GeneID 10155).
Método de purificación Immunogen affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators, Zinc Finger
Primario o secundario Primary
ID de gen (Entrez) 10155
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.