missing translation for 'onlineSavingsMsg'
Learn More

Junctional Cadherin Complex Regulator Antibody, Novus Biologicals™

Código de producto. 18630659 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
25 μL
Tamaño de la unidad:
100 microlitros
25 microlitros
Código de producto. Cantidad unitSize
18630659 25 μL 25 microlitros
18690649 100 μg 100 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18630659

Marca: Novus Biologicals NBP26271925ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Junctional Cadherin Complex Regulator Polyclonal antibody specifically detects Junctional Cadherin Complex Regulator in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Junctional Cadherin Complex Regulator
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulación PBS (pH 7.2) and 40% Glycerol
Alias de gen C11orf63, chromosome 11 open reading frame 63
Especie del huésped Rabbit
Inmunógeno This antibody was developed against a recombinant protein corresponding to amino acids: YAKQVKEYNMKTLSILSKPQTEKTQKKSAIPRQKALEYAKTIPKPKPSNLTHQASKEQKNPTYAGKEESLPEISLLEI
Método de purificación Protein A purified
Cantidad 25 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 79864
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.