missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
IZUMO1 Polyclonal specifically detects IZUMO1 in Human, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.
Especificaciones
Especificaciones
| Antígeno | IZUMO1 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Immunocytochemistry |
| Formulación | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Alias de gen | FLJ61440, IZUMO, izumo sperm-egg fusion 1, izumo sperm-egg fusion protein 1, MGC34799, OBF, Oocyte binding/fusion factor, Sperm-specific protein izumo |
| Símbolos de los genes | IZUMO1 |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against Recombinant Protein corresponding to amino acids:VVLALKSLEKDYLPGHLDAKHHKAMMERVENAVKDFQELSLNEDAYMGVVDEATLQKGSWSLLKDLKRITDSDVKGDLFVKELFWMLHLQKETFATYVARFQKEAYCPNKCGVMLQTLIWCKNCKKEVHACRKSYDCGERNVEVPQM |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?