missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITM2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA549585
Detalles adicionales : Peso : 0.01000kg
Descripción
This gene encodes a type II membrane protein that belongs to the ITM2 family. Studies in mouse suggest that it may be involved in osteo- and chondrogenic differentiation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Especificaciones
ITM2A | |
Polyclonal | |
Unconjugated | |
ITM2A | |
AW108051; BRICD2A; BRICHOS domain containing 2A; E25; E25A; Integral membrane protein 2A; Itm2; ITM2A; Protein E25; UNQ603/PRO1189 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
9452 | |
Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C | |
Liquid |
Flow Cytometry, Western Blot | |
1.4 mg/mL | |
PBS with 0.09% sodium azide | |
O43736 | |
ITM2A | |
KLH conjugated synthetic peptide between 22-49 amino acids from the N-terminal region of human ITM2A. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |