missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITIH4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24069-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The inter-alpha-trypsin inhibitor (ITI, I-alpha-I) family is a classic example of protein-glycosaminoglycoprotein (PGP) complexes. They occur constitutively in plasma at relatively high concentrations and results from alternating combinations of three kinds of heavy chains with a common light chain called the bikunin proteoglycan. Bikunin has two proteinase inhibitor domains and belongs to the Kunitz-type protease inhibitor family; it displays an inhibitory activity against trypsin, leukocyte elastase and plasmin. The heavy chains do not have any protease inhibitory properties but have the capacity to interact in vitro and in vivo with hyaluronic acid and this binding promotes the stability of the extra-cellular matrix. The ITI protein family is suspected of playing a key role in the extra-cellular matrix biology.Especificaciones
| ITIH4 | |
| Polyclonal | |
| Unconjugated | |
| ITIH4 | |
| Gp120, H4P, IHRP, ITI HC4, ITI heavy chain H4, ITIH4, ITIHL1, PK 120, PK120 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16427, 3700 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A6X935, Q14624 | |
| Itih4 | |
| ITIH4 Fusion Protein Ag21275 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido