missing translation for 'onlineSavingsMsg'
Learn More

ISG15/UCRP Antibody (16D8), Novus Biologicals™

Código de producto. 30036100 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
50 μL
100 μL
Tamaño de la unidad:
100 microlitros
50 microlitros
Código de producto. Cantidad unitSize
30036100 100 μL 100 microlitros
30034038 50 μL 50 microlitros
2 options
This item is not returnable. View return policy

Product Code. 30036100

Brand: Novus Biologicals NBP326194100ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ISG15/UCRP Monoclonal antibody specifically detects ISG15/UCRP in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ISG15/UCRP
Aplicaciones Flow Cytometry, ELISA, Immunohistochemistry
Clasificación Monoclonal
Clon 16D8
Conjugado Unconjugated
Dilución Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200
Formulación PBS, pH 7.4, 150mM NaCl and 50% glycerol
Alias de gen G1P2interferon-induced 17-kDa/15-kDa protein, IFI15hUCRP, interferon, alpha-inducible protein (clone IFI-15K), Interferon-induced 15 kDa protein, Interferon-induced 17 kDa protein, interferon-stimulated protein, 15 kDa, ISG15 ubiquitin-like modifier, Ubiquitin cross-reactive protein, ubiquitin-like protein ISG15, UCRPIP17
Especie del huésped Rabbit
Inmunógeno A synthesized peptide derived from Human ISG15/UCRP [UniProt P05161]
Método de purificación Affinity purified
Cantidad 100 μL
Estado normativo RUO
Disciplina de investigación Cell Biology, Signal Transduction, Ubiquitin Proteasome Pathway
Primario o secundario Primary
ID de gen (Entrez) 9636
Especies diana Human
Contenido y almacenamiento Store at -20 to -70C. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.