Learn More
IRP2 Antibody, Novus Biologicals™
Código de producto. 18751454 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18751454 0.025 mL 25 microlitros
18779133 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18751454 Proveedor Novus Biologicals N.º de proveedor NB1001798SS
Rabbit Polyclonal Antibody has been used in 7 publications
IRP2 Polyclonal specifically detects IRP2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Gel Supershift Assay.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno IRP2
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 1.15 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:500-1:1000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunohistochemistry-Paraffin 1:200-1:500, Gel Super Shift Assays reported in scientific literature (PMID 26752519), Gel Supershift Assay
Formulación Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
N.º de referencia del gen Q811J3
Alias de gen ACO3, EC 4.2.1.3, FLJ23381, IRE-BP 2, Iron regulatory protein 2, iron-responsive element binding protein 2, iron-responsive element-binding protein 2, IRP2IRP2AD
Símbolos de los genes IREB2
Especie del huésped Rabbit
Inmunógeno A synthetic peptide made to an internal portion of the murine IRP2 protein sequence (between residues 100-200). [UniProt Q811J3]
Método de purificación Affinity Purified
Cantidad 0.025 mL
Estado normativo RUO
Disciplina de investigación HIF Target Genes, Hypoxia, Lipid and Metabolism
Primario o secundario Primary
ID de gen (Entrez) 3658
Especificidad de la prueba Mouse. Preliminary results have shown poor reactivity against human protein. Immunogen has 94% homology with human, primate and rat proteins.
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Mostrar más Mostrar menos
For Research Use Only