missing translation for 'onlineSavingsMsg'
Learn More

IRF3 Mouse anti-Human, Clone: PCRP-IRF3-1E6, Novus Biologicals™

Código de producto. p-200084322 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18663059 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18663059 Supplier Novus Biologicals Supplier No. NBP307529100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IRF3 Monoclonal specifically detects IRF3 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno IRF3
Aplicaciones Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
Clasificación Monoclonal
Clon PCRP-IRF3-1E6
Concentración 0.2 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL, Protein Array
Formulación 10 mM PBS with 0.05% BSA
Alias de gen interferon regulatory factor 3, IRF-3
Especie del huésped Mouse
Inmunógeno Recombinant full-length human IRF3 protein
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 3661
Especies diana Human
Contenido y almacenamiento Store at 4°C.
Formulario Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.