missing translation for 'onlineSavingsMsg'
Learn More

IRF3 Antibody - BSA Free, Novus Biologicals™

Código de producto. p-200066681 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
Tamaño de la unidad:
25 microgramos
Código de producto. Cantidad unitSize
18522451 0.025 mg 25 microgramos
1 options
This item is not returnable. View return policy

Product Code. 18522451

Brand: Novus Biologicals NBP1765980.025mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IRF3 Polyclonal specifically detects IRF3 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno IRF3
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2 μg/mL
Formulación PBS with 0.02% Sodium Azide
N.º de referencia del gen NP_001562
Alias de gen interferon regulatory factor 3, IRF-3
Símbolos de los genes IRF3
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a peptide corresponding to 14 amino acids near the center of human IRF3. The immunogen is located within amino acids 150 - 200 of IRF3. Amino Acid Squence: CPQPLRSPSLDNPT
Peso molecular del antígeno 50 kDa
Método de purificación Affinity Purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Immunology, Innate Immunity, Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 3661
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.