Learn More
IRE1 alpha, p Ser724 Antibody, HRP, Novus Biologicals™
Código de producto. 18771634 Tienda Bio Techne Productos
Change view
Click to view available options
Tamaño de la unidad:
0.10 ml
Código de producto. unitSize
18771634 0.10 ml
1 options
Código de producto. 18771634
Marca: Novus Biologicals NB1002323H
Rabbit Polyclonal Antibody
IRE1 alpha Polyclonal specifically detects IRE1 alpha in Human, Mouse, Rat, Porcine, Drosophila, Goat, Mammal, Monkey, Primate, Rabbit, Golden Syrian Hamster samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Especificaciones
Especies diana Pig
For Research Use Only