missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
IRAK4 Monoclonal antibody specifically detects IRAK4 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF
Especificaciones
Especificaciones
| Antígeno | IRAK4 |
| Aplicaciones | Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF |
| Clasificación | Monoclonal |
| Clon | 413613 |
| Conjugado | DyLight 755 |
| Dilución | Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready |
| Formulación | 50mM Sodium Borate |
| Alias de gen | EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 |
| Especie del huésped | Rat |
| Inmunógeno | E. coli-derived recombinant human IRAK4, Met1-Ser460, Accession # Q9NWZ3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?