Learn More
Invitrogen™ Quant-iT™ RiboGreen™ RNA Reagent
One of most sensitive fluorescent dyes available for quantitation of RNA in solution
Marca: Invitrogen™ R11491
759.65 EUR hasta el 2024-12-31
Use el código promocional "22915" para aplicar la oferta.
Detalles adicionales : CAS : 9048-46-8 Peso : 0.02000kg
Descripción
- Used with linear fluorescence detection in range of 1-200ng of RNA
- Available in kit complete with buffer and RNA standard (Cat. No. O11490)
DNA and RNA Purification and Analysis, Nucleic Acid Quantitation, RNA Quantitation
Información de órdenes
Shipping Condition: Room temperature
For Research Use Only. Not for use in diagnostic procedures.