missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
INTS8 Polyclonal antibody specifically detects INTS8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | INTS8 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Janelia Fluor 646 |
| Dilución | Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulación | 50mM Sodium Borate |
| Alias de gen | C8orf52, FLJ20530, INT8, integrator complex subunit 8, protein kaonashi-1 |
| Especie del huésped | Rabbit |
| Inmunógeno | The immunogen recognized by this antibody maps to a region between residues 200 and 250 of human FLJ20530/KAONASHI1 using the numbering given in TrEMBL entry Q7Z314 (GeneID 55656). |
| Método de purificación | Immunogen affinity purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?