Learn More
INTS4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16130-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
INTS4 is a subunit of the Integrator complex, which associates with the C-terminal domain of RNA polymerase II large subunit.Especificaciones
| INTS4 | |
| Polyclonal | |
| Unconjugated | |
| INTS4 | |
| INT4, integrator complex subunit 4, INTS4, MST093 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 101861, 308837, 92105 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8CIM8, Q96HW7 | |
| Ints4 | |
| INTS4 Fusion Protein Ag9116 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |