missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Integrin beta 3/CD61 Monoclonal specifically detects Integrin beta 3/CD61 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Especificaciones
Especificaciones
| Antígeno | Integrin beta 3/CD61 |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Clasificación | Monoclonal |
| Clon | F11 |
| Conjugado | DyLight 350 |
| Dilución | Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gen | CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa |
| Símbolos de los genes | ITGB3 |
| Especie del huésped | Mouse |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?