Learn More
Integrin alpha 5/CD49e Antibody (CL6940) [mFluor Violet 610 SE], Novus Biologicals™
Código de producto. 30296494 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30296494 0.1 mL 0.10 ml
1 options
Código de producto. 30296494 Proveedor Novus Biologicals N.º de proveedor NBP276518MFV610
Mouse Monoclonal Antibody
Integrin alpha 5/CD49e Monoclonal antibody specifically detects Integrin alpha 5/CD49e in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Integrin alpha 5/CD49e
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
Clasificación Monoclonal
Clon CL6940
Conjugado mFluor Violet 610 SE
Formulación 50mM Sodium Borate
Alias de gen CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), very late activation protein 5, alpha subunit, VLA-5
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P08648, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación CD Markers, Cell Biology, Cytoskeleton Markers, GPCR, Immunology, Signal Transduction, Stem Cells
Primario o secundario Primary
ID de gen (Entrez) 3678
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1