Learn More
Integrin alpha 4/CD49d Antibody (CL11294), Novus Biologicals™
Código de producto. 18680047 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
25 μg
Tamaño de la unidad:
100 microlitros
25 microlitros
18680047 25 μg 25 microlitros
18674298 100 μg 100 microlitros
2 options
Código de producto. 18680047
Marca: Novus Biologicals NBP30799925ul
Mouse Monoclonal Antibody
Integrin alpha 4/CD49d Monoclonal antibody specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Integrin alpha 4/CD49d
Aplicaciones Western Blot
Clasificación Monoclonal
Clon CL11294
Conjugado Unconjugated
Dilución Western Blot 1 μg/ml
Formulación PBS, pH 7.2, containing 40% glycerol
Alias de gen 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Especie del huésped Mouse
Inmunógeno This antibody was developed against a synthetic peptide corresponding to amino acids: ERDNQWLGVTLSRQPGENGS
Método de purificación Protein A purified
Cantidad 25 μg
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Microautophagy, Neuronal Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 3676
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG2b