Learn More
Integrin alpha 4 beta 7/LPAM-1 Antibody (CL11293) [Alexa Fluor™ 532], Novus Biologicals™
Código de producto. 30294078 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30294078 0.1 mL 0.10 ml
1 options
Código de producto. 30294078
Marca: Novus Biologicals NBP321189AF532
Mouse Monoclonal Antibody
Integrin alpha 4 beta 7/LPAM-1 Monoclonal antibody specifically detects Integrin alpha 4 beta 7/LPAM-1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Integrin alpha 4 beta 7/LPAM-1
Aplicaciones Western Blot
Clasificación Monoclonal
Clon CL11293
Conjugado Alexa Fluor 532
Formulación 50mM Sodium Borate
Alias de gen 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from P13612, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Innate Immunity
Primario o secundario Primary
ID de gen (Entrez) 3676
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1