Learn More
Integrin alpha 3/CD49c Antibody (CL6937) [Janelia Fluor™ 646], Novus Biologicals™
Código de producto. 30292804 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30292804 0.1 mL 0.10 ml
1 options
Código de producto. 30292804
Proveedor: Novus Biologicals NBP276515JF646
Mouse Monoclonal Antibody
Integrin alpha 3/CD49c Monoclonal antibody specifically detects Integrin alpha 3/CD49c in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Integrin alpha 3/CD49c
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
Clasificación Monoclonal
Clon CL6937
Conjugado Janelia Fluor 646
Formulación 50mM Sodium Borate
Alias de gen antigen identified by monoclonal J143, CD49 antigen-like family member C, CD49c, CD49c antigen, FLJ34631, FLJ34704, FRP-2, Galactoprotein B3, GAP-B3, GAPB3CD49C, integrin alpha-3, integrin, alpha 3 (antigen CD49C, alpha 3 subunit of VLA-3 receptor), MSK18, VCA-2, very late activation protein 3 receptor, alpha-3 subunit, VL3A, VLA-3 subunit alpha, VLA3a
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P26006, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 3675
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1